Product Details

SNP ID
rs201921903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55294385 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGAGAGTATAAATGAAGGGATTT[A/G]GTAAAGGGGCAATTGAGGTATAGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR6C6 PubMed Links

Gene Details

Gene
OR6C6
Gene Name
olfactory receptor family 6 subfamily C member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005493.1 848 Missense Mutation CCA,CTA P283L NP_001005493.1

View Full Product Details