Product Details

SNP ID
rs202208466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56229169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCCGCCTGGCCCTTCCAGCAAC[C/G]CTGTTAGTAACGGCAAAGAAACCCG
Phenotype
MIM: 612104 MIM: 608730
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD52 PubMed Links

Gene Details

Gene
ANKRD52
Gene Name
ankyrin repeat domain 52
There are no transcripts associated with this gene.

Gene
NABP2
Gene Name
nucleic acid binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024068.3 736 Missense Mutation CCT,GCT P198A NP_076973.1
XM_005269146.2 736 Missense Mutation CCT,GCT P241A XP_005269203.1
XM_005269147.2 736 Missense Mutation CCT,GCT P214A XP_005269204.1
XM_005269148.4 736 Missense Mutation CCT,GCT P198A XP_005269205.1
XM_005269149.4 736 Missense Mutation CCT,GCT P198A XP_005269206.1
Gene
RNF41
Gene Name
ring finger protein 41
There are no transcripts associated with this gene.

Gene
SLC39A5
Gene Name
solute carrier family 39 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135195.1 736 Intron NP_001128667.1
NM_173596.2 736 Intron NP_775867.2
XM_005268803.1 736 Intron XP_005268860.1
XM_011538198.1 736 Intron XP_011536500.1
XM_011538199.1 736 Intron XP_011536501.1
XM_011538200.1 736 Intron XP_011536502.1
XM_011538201.1 736 Intron XP_011536503.1
XM_017019185.1 736 Intron XP_016874674.1
XM_017019186.1 736 Intron XP_016874675.1
XM_017019187.1 736 Intron XP_016874676.1

View Full Product Details