Product Details
- SNP ID
-
rs201467517
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:49973324 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCTCAGTCATGCGCTGGCCCACA[A/G]CACTTCCCTCCGTGCTACAGATTGC
- Phenotype
-
MIM: 600442
MIM: 601383
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AQP5
PubMed Links
Gene Details
- Gene
- AQP5
- Gene Name
- aquaporin 5
There are no transcripts associated with this gene.
- Gene
- AQP6
- Gene Name
- aquaporin 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001652.3 |
488 |
Missense Mutation |
ACA,GCA |
T51A |
NP_001643.2 |
View Full Product Details