Product Details

SNP ID
rs201229938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:22625321 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCCGTCCCAGCGCCCCCAGCCC[C/T]CCCGCGAGGGCGCCCCGGGACGGAA
Phenotype
MIM: 609858
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ETNK1 PubMed Links

Gene Details

Gene
ETNK1
Gene Name
ethanolamine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039481.1 177 Missense Mutation CCC,CTC P53L NP_001034570.1
NM_018638.4 177 Missense Mutation CCC,CTC P53L NP_061108.2
XM_005253420.4 177 Missense Mutation CCC,CTC P53L XP_005253477.1
XM_017019580.1 177 Missense Mutation CCC,CTC P53L XP_016875069.1
XM_017019581.1 177 Missense Mutation CCC,CTC P53L XP_016875070.1
Gene
LOC105369691
Gene Name
uncharacterized LOC105369691
There are no transcripts associated with this gene.

View Full Product Details