Product Details
- SNP ID
-
rs201561325
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:34943102 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGCAACAGGGACATCGTGGAGAC[A/G]GTGCTCAACCTGGTAAGGAAAAGGC
- Phenotype
-
MIM: 604889
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NBEA
PubMed Links
Gene Details
- Gene
- NBEA
- Gene Name
- neurobeachin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001204197.1 |
816 |
Intron |
|
|
NP_001191126.1 |
NM_015678.4 |
816 |
Silent Mutation |
ACA,ACG |
T94T |
NP_056493.3 |
XM_005266346.4 |
816 |
Silent Mutation |
ACA,ACG |
T94T |
XP_005266403.1 |
XM_005266347.4 |
816 |
Silent Mutation |
ACA,ACG |
T94T |
XP_005266404.2 |
XM_005266348.4 |
816 |
Silent Mutation |
ACA,ACG |
T94T |
XP_005266405.2 |
XM_006719803.3 |
816 |
Silent Mutation |
ACA,ACG |
T94T |
XP_006719866.1 |
XM_006719805.3 |
816 |
Silent Mutation |
ACA,ACG |
T94T |
XP_006719868.1 |
XM_006719806.3 |
816 |
Silent Mutation |
ACA,ACG |
T94T |
XP_006719869.1 |
XM_011535046.2 |
816 |
Intron |
|
|
XP_011533348.1 |
XM_011535047.2 |
816 |
Silent Mutation |
ACA,ACG |
T94T |
XP_011533349.1 |
XM_017020544.1 |
816 |
Silent Mutation |
ACA,ACG |
T94T |
XP_016876033.1 |
XM_017020545.1 |
816 |
Silent Mutation |
ACA,ACG |
T94T |
XP_016876034.1 |
XM_017020546.1 |
816 |
Intron |
|
|
XP_016876035.1 |
View Full Product Details