Product Details

SNP ID
rs201983374
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:20189082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACAAGGCCAGGCGTTGCACTTC[C/A]CCAGCCGCTGCATGGAGAAGCCGTC
Phenotype
MIM: 121011
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
GJB2 PubMed Links

Gene Details

Gene
GJB2
Gene Name
gap junction protein beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004004.5 706 Missense Mutation GGG,GTG G167V NP_003995.2
XM_011535049.2 706 Missense Mutation GGG,GTG G167V XP_011533351.1

View Full Product Details