Product Details

SNP ID
rs202134897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:102767386 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTTGGATGGTGCAGTGGGTAAG[A/G]AATACAAATGAGACCCCGAACAAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC168 PubMed Links

Gene Details

Gene
CCDC168
Gene Name
coiled-coil domain containing 168
There are no transcripts associated with this gene.

Gene
TEX30
Gene Name
testis expressed 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286775.1 645 Intron NP_001273704.1
NM_001286776.1 645 Missense Mutation CCT,TCT P90S NP_001273705.1
NM_138779.4 645 Missense Mutation CCT,TCT P131S NP_620134.3
XM_005254097.2 645 Missense Mutation CCT,TCT P131S XP_005254154.1
XM_011521135.2 645 Missense Mutation CCT,TCT P131S XP_011519437.1
XM_011521136.2 645 Missense Mutation CCT,TCT P87S XP_011519438.1
XM_017020852.1 645 Missense Mutation CCT,TCT P131S XP_016876341.1

View Full Product Details