Product Details

SNP ID
rs201556336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:20404133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGCTGCATTACAAGAAATTCAC[C/T]GGGGCTGCACTTGACTCTTCAACTT
Phenotype
MIM: 609877
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRYL1 PubMed Links

Gene Details

Gene
CRYL1
Gene Name
crystallin lambda 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015974.2 862 Missense Mutation CAG,CGG Q319R NP_057058.2
XM_005266415.3 862 Missense Mutation CAG,CGG Q265R XP_005266472.1
XM_005266416.4 862 Intron XP_005266473.1
XM_017020624.1 862 Missense Mutation CAG,CGG Q234R XP_016876113.1

View Full Product Details