Product Details
- SNP ID
-
hCV190008358
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:58207947 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAATTTTTTACAGCCTGTCAGAGT[C/T]GTTCAGTATAATATCAATACAGAAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACTR10
PubMed Links
Gene Details
- Gene
- ACTR10
- Gene Name
- actin-related protein 10 homolog
There are no transcripts associated with this gene.
View Full Product Details