Product Details
- SNP ID
-
rs199507846
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:24435052 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCAGGCACAGGGCCTTGGCTGAA[C/T]TGAGATGCATCAAACTGTCTGGGAG
- Phenotype
-
MIM: 607519
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PARP4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs62000386] are located under a probe and SNP(s) [rs9511259] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PARP4
- Gene Name
- poly(ADP-ribose) polymerase family member 4
View Full Product Details