Product Details

SNP ID
rs199891204
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:112068797 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCAGCACTACCAGGGCGCGGGCG[C/T]GGGCGTGAACGGCACGGTGCCCCTG
Phenotype
MIM: 602148
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00403 PubMed Links

Gene Details

Gene
LINC00403
Gene Name
long intergenic non-protein coding RNA 403
There are no transcripts associated with this gene.

Gene
SOX1
Gene Name
SRY-box 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005986.2 1199 Missense Mutation GCG,GTG A380V NP_005977.2

View Full Product Details