Product Details

SNP ID
rs199897812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:26759018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAGGTGTAATCCGCTATCAAGGA[A/G]TAGAGGGTGAAAGGCATCCAGCAAG
Phenotype
MIM: 600752
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR12 PubMed Links

Gene Details

Gene
GPR12
Gene Name
G protein-coupled receptor 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005288.3 1032 Silent Mutation TAC,TAT Y270Y NP_005279.1
XM_005266360.3 1032 Silent Mutation TAC,TAT Y111Y XP_005266417.1

View Full Product Details