Product Details

SNP ID
rs199906293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:52844587 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACATTTTCATTGGCTCCCTTCTT[C/T]GGGGACAGGTACCTGCCAGGAGGGG
Phenotype
MIM: 603580
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCDH8 PubMed Links

Gene Details

Gene
PCDH8
Gene Name
protocadherin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002590.3 3390 Silent Mutation CCA,CCG P1062P NP_002581.2
NM_032949.2 3390 Silent Mutation CCA,CCG P965P NP_116567.1

View Full Product Details