Product Details

SNP ID
rs201073774
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24631978 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCACTGTCATCTTCACCTCTTG[C/T]AGTGTGTGTGAGTGTTTTCCCAGGG
Phenotype
MIM: 123910
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GZMB PubMed Links

Gene Details

Gene
GZMB
Gene Name
granzyme B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004131.4 530 Silent Mutation CTA,CTG L160L NP_004122.2
XM_011536685.2 530 Silent Mutation CTA,CTG L148L XP_011534987.1

View Full Product Details