Product Details
- SNP ID
-
rs201617523
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:93184072 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCTACTTTCCTGTTCTCATCCCT[C/T]CTGAACATCCTTCCAAGCAGTCTGT
- Phenotype
-
MIM: 609485
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MOAP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs547420517] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MOAP1
- Gene Name
- modulator of apoptosis 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022151.4 |
541 |
Silent Mutation |
AGA,AGG |
R57R |
NP_071434.2 |
- Gene
- TMEM251
- Gene Name
- transmembrane protein 251
View Full Product Details