Product Details

SNP ID
rs199821940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:93186507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTGGACAGTTATTTTTCTGGTA[C/G]CTTACTTACAGATGTTTTTGTTCCT
Phenotype
MIM: 609485
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MOAP1 PubMed Links

Gene Details

Gene
MOAP1
Gene Name
modulator of apoptosis 1
There are no transcripts associated with this gene.

Gene
TMEM251
Gene Name
transmembrane protein 251
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098621.1 704 Missense Mutation CCT,GCT P116A NP_001092091.1
NM_015676.1 704 Missense Mutation NP_056491.1

View Full Product Details