Product Details

SNP ID
rs201701584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:70522696 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGACAGGTAACGCAACTTTCCCA[A/T]CACATTTAATCCTTCCATGTTGTTC
Phenotype
MIM: 603712
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ADAM20 PubMed Links

Gene Details

Gene
ADAM20
Gene Name
ADAM metallopeptidase domain 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003814.4 11353 Missense Mutation ATG,TTG M738L NP_003805.3
XM_005268151.3 11353 Missense Mutation ATG,TTG M738L XP_005268208.1

View Full Product Details