Product Details

SNP ID
rs200194901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24148324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAACCAGATGGGTTGAGCTTCCCC[C/G]AAGGGCAGGAGGAGCCAGATGAGCA
Phenotype
MIM: 600654 MIM: 602161 MIM: 612487
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EMC9 PubMed Links

Gene Details

Gene
EMC9
Gene Name
ER membrane protein complex subunit 9
There are no transcripts associated with this gene.

Gene
MIR7703
Gene Name
microRNA 7703
There are no transcripts associated with this gene.

Gene
PSME1
Gene Name
proteasome activator subunit 1
There are no transcripts associated with this gene.

Gene
PSME2
Gene Name
proteasome activator subunit 2
There are no transcripts associated with this gene.

Gene
RNF31
Gene Name
ring finger protein 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001310332.1 247 UTR 5 NP_001297261.1
NM_017999.4 247 Missense Mutation CAA,GAA Q136E NP_060469.4

View Full Product Details