Product Details

SNP ID
rs200985291
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:98716279 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGGAACAAAGCCGATGATCACA[C/T]GCCCATCAGTGAGCCAACTTGGTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C14orf177 PubMed Links

Gene Details

Gene
C14orf177
Gene Name
chromosome 14 open reading frame 177
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182560.2 507 Missense Mutation CGC,TGC R30C NP_872366.2

View Full Product Details