Product Details

SNP ID
rs200553632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20198325 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTAACCCTGTGATATATAGTCT[G/T]AGGAACAAAGATATGAGAAAAGCTC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR11G2 PubMed Links

Gene Details

Gene
OR11G2
Gene Name
olfactory receptor family 11 subfamily G member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005503.1 990 Silent Mutation CTG,CTT L330L NP_001005503.1

View Full Product Details