Product Details

SNP ID
rs201384449
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21288026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGAGACTTGCCAGTTAGAGACA[C/T]AGATGCTATACCTCTGGTGCTACCA
Phenotype
MIM: 605446
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPGRIP1 PubMed Links

Gene Details

Gene
RPGRIP1
Gene Name
retinitis pigmentosa GTPase regulator interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020366.3 50 Missense Mutation ACA,ATA T17I NP_065099.3
XM_005267879.2 50 Intron XP_005267936.1
XM_005267880.2 50 Intron XP_005267937.1
XM_005267881.3 50 Intron XP_005267938.1
XM_011536978.1 50 Intron XP_011535280.1
XM_011536979.1 50 Intron XP_011535281.1
XM_011536980.1 50 Intron XP_011535282.1
XM_011536981.1 50 Intron XP_011535283.1
XM_011536982.1 50 Intron XP_011535284.1
XM_017021473.1 50 Intron XP_016876962.1

View Full Product Details