Product Details

SNP ID
rs201385630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21090674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGGGACGCGTCAGCCTGGGGTGG[A/C]CGGCGGCCCCTAGCCCGGCGGCTGT
Phenotype
MIM: 610018 MIM: 605036
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARHGEF40 PubMed Links

Gene Details

Gene
ARHGEF40
Gene Name
Rho guanine nucleotide exchange factor 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278529.1 2199 Intron NP_001265458.1
NM_001278530.1 2199 Intron NP_001265459.1
NM_018071.4 2199 Intron NP_060541.3
XM_005267844.2 2199 Intron XP_005267901.1
XM_011536937.2 2199 Intron XP_011535239.1
XM_011536938.2 2199 Intron XP_011535240.1
XM_017021434.1 2199 Intron XP_016876923.1
XM_017021435.1 2199 Intron XP_016876924.1
XM_017021436.1 2199 Intron XP_016876925.1
XM_017021437.1 2199 Intron XP_016876926.1
Gene
TMEM253
Gene Name
transmembrane protein 253
There are no transcripts associated with this gene.

Gene
ZNF219
Gene Name
zinc finger protein 219
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101672.1 2199 Silent Mutation CGG,CGT R677R NP_001095142.1
NM_001102454.1 2199 Silent Mutation CGG,CGT R677R NP_001095924.1
NM_016423.2 2199 Silent Mutation CGG,CGT R677R NP_057507.2
XM_005267739.1 2199 Silent Mutation CGG,CGT R723R XP_005267796.1
XM_006720163.2 2199 Silent Mutation CGG,CGT R677R XP_006720226.1
XM_006720164.3 2199 Silent Mutation CGG,CGT R677R XP_006720227.1
XM_011536825.1 2199 Silent Mutation CGG,CGT R714R XP_011535127.1
XM_017021354.1 2199 Silent Mutation CGG,CGT R677R XP_016876843.1
XM_017021355.1 2199 Silent Mutation CGG,CGT R677R XP_016876844.1

View Full Product Details