Product Details

SNP ID
rs199766168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22875602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTAGCCTCAGTCTCCCACCCCCA[C/G]TCCTGCCATTGGCTGCTGGACCCCC
Phenotype
MIM: 609921 MIM: 616955
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LRP10 PubMed Links

Gene Details

Gene
LRP10
Gene Name
LDL receptor related protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005267510.1 1369 Missense Mutation CAC,CAG H226Q XP_005267567.1
Gene
REM2
Gene Name
RRAD and GEM like GTPase 2
There are no transcripts associated with this gene.

View Full Product Details