Product Details

SNP ID
rs200199139
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24268390 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCTGAATAGCTGCTCGTCTGTC[A/G]TGGAGTCCCGGCACCAGCCCTCCTG
Phenotype
MIM: 601905 MIM: 190195
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RABGGTA PubMed Links

Gene Details

Gene
RABGGTA
Gene Name
Rab geranylgeranyltransferase alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004581.5 1549 Missense Mutation ACG,ATG T346M NP_004572.3
NM_182836.2 1549 Missense Mutation ACG,ATG T346M NP_878256.1
Gene
TGM1
Gene Name
transglutaminase 1
There are no transcripts associated with this gene.

View Full Product Details