Product Details

SNP ID
rs201899051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:57566487 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTGCCTGTACCTGCATTTCCAG[A/G]AACGCTGAGCACTGTCCCAATGGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC35F4 PubMed Links

Gene Details

Gene
SLC35F4
Gene Name
solute carrier family 35 member F4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206920.1 1518 Missense Mutation CCT,TCT P401S NP_001193849.1
NM_001306087.1 1518 Missense Mutation CCT,TCT P402S NP_001293016.1
XM_011536720.2 1518 Missense Mutation CCT,TCT P449S XP_011535022.1
XM_011536721.2 1518 Missense Mutation CCT,TCT P448S XP_011535023.1
XM_011536723.2 1518 Missense Mutation CCT,TCT P389S XP_011535025.1
XM_011536724.2 1518 Missense Mutation CCT,TCT P384S XP_011535026.1
XM_011536725.1 1518 Missense Mutation CCT,TCT P279S XP_011535027.1
XM_017021258.1 1518 Missense Mutation CCT,TCT P449S XP_016876747.1
XM_017021259.1 1518 Intron XP_016876748.1
XM_017021260.1 1518 Missense Mutation CCT,TCT P279S XP_016876749.1
XM_017021261.1 1518 Intron XP_016876750.1

View Full Product Details