Product Details

SNP ID
rs200773885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:103913970 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGCAAAAATCTTACCTTTCAAAG[C/T]CTTACTTCTTTTATCTAAAATAAAA
Phenotype
MIM: 604573
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C14orf2 PubMed Links

Gene Details

Gene
C14orf2
Gene Name
chromosome 14 open reading frame 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127393.1 236 Missense Mutation ACT,GCT T64A NP_001120865.1
NM_004894.2 236 Missense Mutation ACT,GCT T47A NP_004885.1

View Full Product Details