Product Details

SNP ID
rs201173826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24265727 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGAGGAGGACCAGCCTGGGGCAG[A/G]AGGCAAGAGGCTGGAGCACTGCAGG
Phenotype
MIM: 601905 MIM: 190195
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RABGGTA PubMed Links

Gene Details

Gene
RABGGTA
Gene Name
Rab geranylgeranyltransferase alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004581.5 2104 Missense Mutation TCC,TTC S531F NP_004572.3
NM_182836.2 2104 Missense Mutation TCC,TTC S531F NP_878256.1
Gene
TGM1
Gene Name
transglutaminase 1
There are no transcripts associated with this gene.

View Full Product Details