Product Details
- SNP ID
-
rs201358563
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:24081247 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGCTCTGAACGGCTCAGAGGAAGA[A/G]GTGGGAGGGGTCCCCGGACCCGGGG
- Phenotype
-
MIM: 605688
MIM: 162080
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CPNE6
PubMed Links
Gene Details
- Gene
- CPNE6
- Gene Name
- copine 6
There are no transcripts associated with this gene.
- Gene
- NRL
- Gene Name
- neural retina leucine zipper
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006177.3 |
618 |
Missense Mutation |
CTC,TTC |
L235F |
NP_006168.1 |
XM_005267708.4 |
618 |
Missense Mutation |
CTC,TTC |
L235F |
XP_005267765.1 |
XM_005267709.3 |
618 |
Missense Mutation |
CTC,TTC |
L235F |
XP_005267766.1 |
XM_005267710.3 |
618 |
Missense Mutation |
CTC,TTC |
L235F |
XP_005267767.1 |
XM_011536801.2 |
618 |
Missense Mutation |
CTC,TTC |
L337F |
XP_011535103.2 |
XM_011536802.1 |
618 |
Missense Mutation |
CTC,TTC |
L235F |
XP_011535104.1 |
XM_011536804.2 |
618 |
Missense Mutation |
CTC,TTC |
L235F |
XP_011535106.1 |
XM_011536805.2 |
618 |
Missense Mutation |
CTC,TTC |
L235F |
XP_011535107.1 |
XM_011536806.2 |
618 |
Missense Mutation |
CTC,TTC |
L232F |
XP_011535108.2 |
XM_017021350.1 |
618 |
Missense Mutation |
CTC,TTC |
L130F |
XP_016876839.1 |
View Full Product Details