Product Details

SNP ID
rs200455244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20460266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGACAGTTACAGGGGCATCGAAC[A/G]TATTTTTTCCCTGGGGGTGCATTCT
Phenotype
MIM: 107748 MIM: 610107 MIM: 164050 MIM: 609865
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APEX1 PubMed Links

Gene Details

Gene
APEX1
Gene Name
apurinic/apyrimidinic endodeoxyribonuclease 1
There are no transcripts associated with this gene.

Gene
OSGEP
Gene Name
O-sialoglycoprotein endopeptidase
There are no transcripts associated with this gene.

Gene
PNP
Gene Name
purine nucleoside phosphorylase
There are no transcripts associated with this gene.

Gene
TMEM55B
Gene Name
transmembrane protein 55B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100814.2 674 Silent Mutation TAC,TAT Y129Y NP_001094284.1
NM_144568.3 674 Silent Mutation TAC,TAT Y122Y NP_653169.2

View Full Product Details