Product Details

SNP ID
rs200710673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22885000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTCTCCAGGGAGACAGTGCTCAC[A/G]AACCGGAGAACCCAGGTATTTGGGG
Phenotype
MIM: 609921 MIM: 616955
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRP10 PubMed Links

Gene Details

Gene
LRP10
Gene Name
LDL receptor related protein 10
There are no transcripts associated with this gene.

Gene
REM2
Gene Name
RRAD and GEM like GTPase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173527.2 1702 Missense Mutation AAA,GAA K144E NP_775798.2
XM_005267383.3 1702 Missense Mutation AAA,GAA K144E XP_005267440.1
XM_011536518.2 1702 Missense Mutation AAA,GAA K176E XP_011534820.1
XM_017021058.1 1702 Missense Mutation AAA,GAA K176E XP_016876547.1
XM_017021059.1 1702 Missense Mutation AAA,GAA K176E XP_016876548.1

View Full Product Details