Product Details
- SNP ID
-
rs202082945
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:46651731 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCGGTAACAGCGAGCTGGACGGCG[C/T]CCCATGGCGACACAGTAACGTCGGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RPL10L
PubMed Links
Gene Details
- Gene
- RPL10L
- Gene Name
- ribosomal protein L10 like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080746.2 |
|
Intron |
|
|
NP_542784.1 |
View Full Product Details