Product Details

SNP ID
rs202167544
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:49619084 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACTTCTTACAGAAGGTTCTTCG[C/G]GTTTTAGGTACGTTGACCATCTTTG
Phenotype
MIM: 612517 MIM: 609193 MIM: 602616 MIM: 180469
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
DNAAF2 PubMed Links
Additional Information
For this assay, SNP(s) [rs17121614] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
DNAAF2
Gene Name
dynein axonemal assembly factor 2
There are no transcripts associated with this gene.

Gene
LRR1
Gene Name
leucine rich repeat protein 1
There are no transcripts associated with this gene.

Gene
MGAT2
Gene Name
mannosyl (alpha-1,6-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase
There are no transcripts associated with this gene.

Gene
RPL36AL
Gene Name
ribosomal protein L36a like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001.4 Intron NP_000992.1

View Full Product Details