Product Details

SNP ID
rs202041575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41313323 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTTTGAATATCCATCTCTGATG[C/T]ATTTACTATGGCTTTTGTTTTTAAG
Phenotype
MIM: 606020
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OIP5 PubMed Links

Gene Details

Gene
OIP5
Gene Name
Opa interacting protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317860.1 604 Missense Mutation ACA,GCA T141A NP_001304789.1
NM_007280.1 604 Missense Mutation ACA,GCA T182A NP_009211.1

View Full Product Details