Product Details

SNP ID
rs201340108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43276472 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGGTCTCAGGGAGCCCCAGCCA[C/T]AGTGACGAAGATGTCCTTGTAGCCT
Phenotype
MIM: 603805 MIM: 606776
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TGM5 PubMed Links

Gene Details

Gene
TGM5
Gene Name
transglutaminase 5
There are no transcripts associated with this gene.

Gene
TGM7
Gene Name
transglutaminase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052955.2 2119 Missense Mutation ATG,GTG M706V NP_443187.1
XM_017021903.1 2119 Missense Mutation ATG,GTG M707V XP_016877392.1

View Full Product Details