Product Details

SNP ID
rs201676094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:52551988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGCTTCCGTTGAGGAAGGTCTTG[C/T]GGAGTGGGAATGTGGTCACCAGTGA
Phenotype
MIM: 605487
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARPP19 PubMed Links

Gene Details

Gene
ARPP19
Gene Name
cAMP regulated phosphoprotein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306191.1 583 Silent Mutation CCA,CCG P114P NP_001293120.1
NM_001306195.1 583 Intron NP_001293124.1
NM_001306196.1 583 Silent Mutation CCA,CCG P114P NP_001293125.1
NM_006628.5 583 Silent Mutation CCA,CCG P95P NP_006619.1
XM_017021868.1 583 Intron XP_016877357.1
XM_017021869.1 583 Silent Mutation CCA,CCG P95P XP_016877358.1
XM_017021870.1 583 Intron XP_016877359.1
XM_017021871.1 583 Silent Mutation CCA,CCG P79P XP_016877360.1

View Full Product Details