Product Details

SNP ID
rs201831894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:49860330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGGGGGTGGATTTTTAGCTCGC[A/G]TATTTTTTCCAGATGTGATAAGCTC
Phenotype
MIM: 609123
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP8B4 PubMed Links

Gene Details

Gene
ATP8B4
Gene Name
ATPase phospholipid transporting 8B4 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024837.3 3796 Missense Mutation ACG,ATG T1148M NP_079113.2
XM_011522046.2 3796 Missense Mutation ACG,ATG T1213M XP_011520348.1
XM_011522047.2 3796 Missense Mutation ACG,ATG T1176M XP_011520349.1
XM_011522048.1 3796 Missense Mutation ACG,ATG T1176M XP_011520350.1
XM_011522049.2 3796 Missense Mutation ACG,ATG T1176M XP_011520351.1
XM_011522051.2 3796 Missense Mutation ACG,ATG T1176M XP_011520353.1
XM_011522052.2 3796 Missense Mutation ACG,ATG T1176M XP_011520354.1
XM_011522053.1 3796 Missense Mutation ACG,ATG T1176M XP_011520355.1
XM_011522056.2 3796 Missense Mutation ACG,ATG T1278M XP_011520358.2
XM_011522058.2 3796 Missense Mutation ACG,ATG T1067M XP_011520360.1
XM_011522059.1 3796 Missense Mutation ACG,ATG T1061M XP_011520361.1
XM_011522060.1 3796 Missense Mutation ACG,ATG T1049M XP_011520362.1
XM_011522061.1 3796 Missense Mutation ACG,ATG T1049M XP_011520363.1
XM_011522062.1 3796 Missense Mutation ACG,ATG T1049M XP_011520364.1
XM_011522063.1 3796 Missense Mutation ACG,ATG T1049M XP_011520365.1
XM_011522064.1 3796 Missense Mutation ACG,ATG T964M XP_011520366.1
XM_011522069.2 3796 Missense Mutation ACG,ATG T893M XP_011520371.1
XM_011522070.1 3796 Missense Mutation ACG,ATG T695M XP_011520372.1
XM_017022587.1 3796 Missense Mutation ACG,ATG T1250M XP_016878076.1
XM_017022588.1 3796 Missense Mutation ACG,ATG T1225M XP_016878077.1
XM_017022589.1 3796 Missense Mutation ACG,ATG T1215M XP_016878078.1
XM_017022590.1 3796 Missense Mutation ACG,ATG T1176M XP_016878079.1
XM_017022591.1 3796 Missense Mutation ACG,ATG T1176M XP_016878080.1
XM_017022592.1 3796 Missense Mutation ACG,ATG T1148M XP_016878081.1
XM_017022593.1 3796 Intron XP_016878082.1
XM_017022594.1 3796 Missense Mutation ACG,ATG T1067M XP_016878083.1
XM_017022595.1 3796 Missense Mutation ACG,ATG T1049M XP_016878084.1
XM_017022596.1 3796 Missense Mutation ACG,ATG T937M XP_016878085.1
XM_017022597.1 3796 Intron XP_016878086.1
XM_017022598.1 3796 Intron XP_016878087.1
XM_017022599.1 3796 Intron XP_016878088.1

View Full Product Details