Product Details

SNP ID
rs201354061
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:38698353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATTACAAAGTTTGACTCCAAGAG[A/T]AGCCCAGAAGTGGCTTCCAGCCCCT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C15orf53 PubMed Links

Gene Details

Gene
C15orf53
Gene Name
chromosome 15 open reading frame 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207444.2 358 Missense Mutation AGA,AGT R116S NP_997327.1

View Full Product Details