Product Details

SNP ID
rs201461230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:81002531 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGACACCATCATCGCGCGCACCA[A/C]GGGGCTCTCCATCCTCACCCACGAC
Phenotype
MIM: 615466
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MESDC1 PubMed Links

Gene Details

Gene
MESDC1
Gene Name
mesoderm development candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022566.2 1578 Missense Mutation AAG,ACG K87T NP_072088.1
Gene
MIR4514
Gene Name
microRNA 4514
There are no transcripts associated with this gene.

View Full Product Details