Product Details

SNP ID
rs199778021
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43203208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTGTTGCCATGGCGGTGAGTCT[A/G]AGGAAGGTGTTCTCGGGTGGGTTTC
Phenotype
MIM: 607089 MIM: 177070
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCNDBP1 PubMed Links

Gene Details

Gene
CCNDBP1
Gene Name
cyclin D1 binding protein 1
There are no transcripts associated with this gene.

Gene
EPB42
Gene Name
erythrocyte membrane protein band 4.2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000119.2 1718 Silent Mutation CTC,CTT L592L NP_000110.2
NM_001114134.1 1718 Silent Mutation CTC,CTT L562L NP_001107606.1
XM_005254225.1 1718 Silent Mutation CTC,CTT L527L XP_005254282.1
XM_011521349.2 1718 Silent Mutation CTC,CTT L592L XP_011519651.1
XM_011521350.2 1718 Silent Mutation CTC,CTT L592L XP_011519652.1
XM_011521351.2 1718 Silent Mutation CTC,CTT L592L XP_011519653.1
XM_011521352.2 1718 Silent Mutation CTC,CTT L580L XP_011519654.1
XM_011521353.2 1718 Silent Mutation CTC,CTT L557L XP_011519655.1
XM_011521354.2 1718 Silent Mutation CTC,CTT L407L XP_011519656.1

View Full Product Details