Product Details

SNP ID
rs201048326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:80948850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTTCCGGCCCATCTGGGTGACA[A/C]TGGACACTGAGGATCACAAAGCCAA
Phenotype
MIM: 608366 MIM: 607783
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CEMIP PubMed Links

Gene Details

Gene
CEMIP
Gene Name
cell migration inducing hyaluronan binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293298.1 4300 Missense Mutation ATG,CTG M1338L NP_001280227.1
NM_001293304.1 4300 Missense Mutation ATG,CTG M1338L NP_001280233.1
NM_018689.2 4300 Missense Mutation ATG,CTG M1338L NP_061159.1
Gene
MESDC2
Gene Name
mesoderm development candidate 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015154.2 4300 Intron NP_055969.1

View Full Product Details