Product Details

SNP ID
rs200523545
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43521509 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTGAGTTCTCCGAGTATGTCTC[G/T]GAGACTGTGGACGTGCCATCCCCAT
Phenotype
MIM: 600178
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
MAP1A PubMed Links

Gene Details

Gene
MAP1A
Gene Name
microtubule associated protein 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002373.5 793 Silent Mutation TCG,TCT S12S NP_002364.5
XM_017022189.1 793 Silent Mutation TCG,TCT S250S XP_016877678.1

View Full Product Details