Product Details

SNP ID
rs199544667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75206271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCCATACCCACCCTTGTACTCT[A/G]CCGGTATGGCAGGACCCCCACTTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C15orf39 PubMed Links

Gene Details

Gene
C15orf39
Gene Name
chromosome 15 open reading frame 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015492.4 453 Missense Mutation ACC,GCC T75A NP_056307.2

View Full Product Details