Product Details

SNP ID
rs199901645
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:289544 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGGGTTCCCCGCAGAAGTAGTA[C/T]GCCACAACGATGCTGTCACACGGCT
Phenotype
MIM: 602844 MIM: 603816 MIM: 608012
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGDIG PubMed Links

Gene Details

Gene
ARHGDIG
Gene Name
Rho GDP dissociation inhibitor gamma
There are no transcripts associated with this gene.

Gene
AXIN1
Gene Name
axin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003502.3 2758 Silent Mutation NP_003493.1
NM_181050.2 2758 Silent Mutation NP_851393.1
XM_011522682.2 2758 Silent Mutation XP_011520984.1
XM_011522683.2 2758 Silent Mutation XP_011520985.1
XM_011522684.2 2758 Silent Mutation XP_011520986.1
XM_011522686.1 2758 Silent Mutation XP_011520988.1
XM_017023743.1 2758 Silent Mutation XP_016879232.1
XM_017023744.1 2758 Silent Mutation XP_016879233.1
XM_017023745.1 2758 Silent Mutation XP_016879234.1
XM_017023746.1 2758 Silent Mutation XP_016879235.1
XM_017023747.1 2758 Intron XP_016879236.1
XM_017023748.1 2758 Intron XP_016879237.1
Gene
PDIA2
Gene Name
protein disulfide isomerase family A member 2
There are no transcripts associated with this gene.

View Full Product Details