Product Details

SNP ID
rs200787815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2757902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTTTGATCCTCAGCGTCGTGCC[C/G]GAGAAGCTAAACAACCAGCTCCTGA
Phenotype
MIM: 606032
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SRRM2 PubMed Links

Gene Details

Gene
SRRM2
Gene Name
serine/arginine repetitive matrix 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016333.3 999 Missense Mutation CGA,GGA R158G NP_057417.3
XM_005255226.1 999 Missense Mutation CGA,GGA R158G XP_005255283.1
XM_006720874.3 999 Missense Mutation CGA,GGA R158G XP_006720937.1
Gene
SRRM2-AS1
Gene Name
SRRM2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details