Product Details
- SNP ID
-
rs201557051
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:89179441 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCTCTGCCTGTCTTTGGGGGTTC[C/T]TGGATGGAGGAGGCCCACCACCCTG
- Phenotype
-
MIM: 114019
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDH15
PubMed Links
Gene Details
- Gene
- CDH15
- Gene Name
- cadherin 15
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004933.2 |
145 |
Missense Mutation |
CCT,CTT |
P23L |
NP_004924.1 |
View Full Product Details