Product Details

SNP ID
rs201557537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:72056233 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGGACTCAGGCAATGATGTCAC[A/G]GATATCGCAGGTCAGTCTTTGGTTG
Phenotype
MIM: 140100 MIM: 140210
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HP PubMed Links

Gene Details

Gene
HP
Gene Name
haptoglobin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126102.2 139 Silent Mutation ACA,ACG T26T NP_001119574.1
NM_001318138.1 139 Silent Mutation ACA,ACG T26T NP_001305067.1
NM_005143.4 139 Silent Mutation ACA,ACG T26T NP_005134.1
Gene
HPR
Gene Name
haptoglobin-related protein
There are no transcripts associated with this gene.

Gene
TXNL4B
Gene Name
thioredoxin like 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142317.1 139 Intron NP_001135789.1
NM_001142318.1 139 Intron NP_001135790.1
NM_001324354.1 139 Intron NP_001311283.1
NM_001324355.1 139 Intron NP_001311284.1
NM_017853.2 139 Intron NP_060323.1
XM_017023377.1 139 Intron XP_016878866.1

View Full Product Details