Product Details

SNP ID
rs201282006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56963047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCAAGGTGATCCAGACCGCCTT[C/G]CAGCGAGCCAGCTACCCAGATATCA
Phenotype
MIM: 118470
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CETP PubMed Links

Gene Details

Gene
CETP
Gene Name
cholesteryl ester transfer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000078.2 173 Missense Mutation TTC,TTG F52L NP_000069.2
NM_001286085.1 173 Missense Mutation TTC,TTG F52L NP_001273014.1
XM_006721124.3 173 Missense Mutation TTC,TTG F52L XP_006721187.1

View Full Product Details