Product Details

SNP ID
rs200238995
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:74302264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGACCTAGGGCTGCCTACAGAAG[C/T]GTACATTTCAGTGGAAGAAGTCCAT
Phenotype
MIM: 157970
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PSMD7 PubMed Links

Gene Details

Gene
PSMD7
Gene Name
proteasome 26S subunit, non-ATPase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002811.4 550 Missense Mutation GCG,GTG A137V NP_002802.2

View Full Product Details