Product Details

SNP ID
rs200473396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30707188 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTGGCTCTCCCTGATTAGGAGGC[C/T]GAGATCGAGACTCGGATTGCTGAGC
Phenotype
MIM: 611421
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC730183 PubMed Links

Gene Details

Gene
LOC730183
Gene Name
uncharacterized LOC730183
There are no transcripts associated with this gene.

Gene
SNORA30
Gene Name
small nucleolar RNA, H/ACA box 30
There are no transcripts associated with this gene.

Gene
SRCAP
Gene Name
Snf2-related CREBBP activator protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006662.2 697 Silent Mutation GCC,GCT A104A NP_006653.2

View Full Product Details