Product Details

SNP ID
rs201287666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:27461503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCACCACGGGTGTAGAGAAGAG[C/T]GAGACAGGCCTTGGCTTTCTCAGCC
Phenotype
MIM: 603246 MIM: 605383
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GTF3C1 PubMed Links

Gene Details

Gene
GTF3C1
Gene Name
general transcription factor IIIC subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286242.1 6024 Silent Mutation NP_001273171.1
NM_001520.3 6024 Silent Mutation NP_001511.2
XM_017023188.1 6024 Silent Mutation XP_016878677.1
Gene
IL21R
Gene Name
interleukin 21 receptor
There are no transcripts associated with this gene.

Gene
IL21R-AS1
Gene Name
IL21R antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details